Anti-CBFA2T2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11336-1-AP-150UL
10084-118EA 402 USD
10084-118
Anti-CBFA2T2 Rabbit Polyclonal Antibody
Antibodies
Core-binding factor, runt domain, alpha subunit 2, translocated to 2(CBFA2T2) belongs to a family of ubiquitously expressed transcriptional repressors that interact with transcription factors bound to promoters of target genes and with histone deacetylases (HDACs) critical for enforcement of repressor function.CBFA2T2, also named as MTGR1 or EHT, is an 85-kDa phosphoprotein. It can bind AML1-MTG8 fusion protein in acute myeloid leukemia. The binding leads to an enhancement of cell proliferation mediated in a murine myeloid model.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: CBFA2T2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR