Anti-ATRIP Rabbit Polyclonal Antibody

Supplier: Proteintech
11327-1-AP-150UL
10083-398EA 402 USD
10083-398
Anti-ATRIP Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:500-1:5000; IP: MCF-7 Cells, 1:500-1:5000

Type: Primary
Antigen: ATRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR