Anti-LAT Rabbit Polyclonal Antibody

Supplier: Proteintech
11326-1-AP-150UL
10089-530EA 402 USD
10089-530
Anti-LAT Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:1000

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR