Anti-NASP Rabbit Polyclonal Antibody

Supplier: Proteintech
11323-1-AP-150UL
10090-826EA 402 USD
10090-826
Anti-NASP Rabbit Polyclonal Antibody
Antibodies
NASP (nuclear autoantigenic sperm protein) is associated with DNA replication, cell proliferation and cell cycle progression through functioning as a Histone H1 binding protein that mediates histone transport to the nucleus. NASP had two isoforms, tNASP (testis type) and sNASP (somatic type). The tNASP was mainly expressed in testis, a variety of malignant tumors, stem cells and embryonic tissues, while sNASP existed in all somatic mitosis cells. This antibody can recognize both isoforms.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Testis, 1:20-1:200; IF: MCF-7, 1:10-1:100

Type: Primary
Antigen: NASP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR