Anti-CD7 Rabbit Polyclonal Antibody

Supplier: Proteintech
11320-1-AP-150UL
10084-378EA 402 USD
10084-378
Anti-CD7 Rabbit Polyclonal Antibody
Antibodies
CD7 is a 40 kD type I transmembrane glycoprotein expressed on thymocytes and mature T cells. It is the most reliable clinical marker of T-cell acute lymphocytic leukemia.

Western Blot: HL-60 Cells, 1:200-1:1000; IHC: Human tonsil Tissue, 1:20-1:200

Type: Primary
Antigen: CD7
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR