Anti-STMN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11311-1-AP-150UL
10095-332EA 402 USD
10095-332
Anti-STMN3 Rabbit Polyclonal Antibody
Antibodies
Stathmin family phosphoproteins participate in the control of microtubule dynamics and have been proposed to be involved in the control of neuronal differentiation. Neuron specific Stathmin-3 (STMN3) is a novel STAT3 (signal transducer and activator of transcription 3) interacting protein. STAT3 is a key contributor to cancer cell migration and invasion. STMN3 exhibits microtubule-destabilizing activity, which is antagonized by STAT3. SCG10 is a widely studied and recognized neuronal differentiation marker, STMN3 is a SCG10-like protein (SCLIP) and is present from the earliest stages of hippocampal neuron differentiation in culture at vesicle-like structures following dynamic microtubules. RNAi mediated inhibition of this gene resulted in increased axonal branching.

Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human Colon cancer Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:1000-1:10000

Type: Primary
Antigen: STMN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR