Anti-RAB18 Rabbit Polyclonal Antibody

Supplier: Proteintech
11304-1-AP-150UL
10093-138EA 402 USD
10093-138
Anti-RAB18 Rabbit Polyclonal Antibody
Antibodies
RAB18 is a member of a family of Ras-related small GTPases. RAB18 plays a role in apical endocytosis/recycling, and may be implicated in transport between the plasma membrane and early endosomes. It also plays a key role in eye and brain development and neurodegeneration. Defects in RAB18 are associated with Warburg micro syndrome type 3.

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RAB18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR