Anti-EIF3I Rabbit Polyclonal Antibody

Supplier: Proteintech
11287-1-AP-150UL
10086-380EA 537.7 USD
10086-380
Anti-EIF3I Rabbit Polyclonal Antibody
Antibodies
Eukaryotic initiation factor (EIF3) complex is the largest of the eIFs and consists of more than 10 nonidentical subunit, and is required for several steps in the initiation of protein synthesis. It associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). Also it can stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of posttermination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. EIF3I is one subunit of EIF3 complex.

IHC: Human breast cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HEK-293 Cells, 1:500-1:5000; Western Blot: HEK-293 Cells, 1:500-1:5000; FC: HeLa Cells, N/A

Type: Primary
Antigen: EIF3I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR