Anti-RNF156 Rabbit Polyclonal Antibody

Supplier: Proteintech
11285-1-AP-150UL
10093-792EA 402 USD
10093-792
Anti-RNF156 Rabbit Polyclonal Antibody
Antibodies
MGRN1(E3 ubiquitin-protein ligase) mediates monoubiquitination at multiple sites of TSG101 in the presence of UBE2D1, but not of UBE2G1, nor UBE2H. It also plays a role in the regulation of endosome-to-lysosome trafficking. Human and mouse mahogunin exhibit a similar expression pattern across multiple tissues with particularly high levels in brain.It has 4 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RNF156
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR