Anti-TMPRSS4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11283-1-AP-150UL
10096-100EA 402 USD
10096-100
Anti-TMPRSS4 Rabbit Polyclonal Antibody
Antibodies
TMPRSS4, also named as CAPH2 and MT-SP2, belongs to the peptidase S1 family. TMPRSS4 is a novel type II transmembrane serine protease that is highly expressed on the cell surface in pancreatic, thyroid and other cancer tissues. It promotes invasion, migration and metastasis of human tumor cells by facilitating an epithelial-mesenchymal transition(EMT). TMPRSS4 is not expressed in normal pancreas or in chronic pancreatitis tissue samples, overexpression appeared to be cancer-specific.

Western Blot:BxPC-3 Cells, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TMPRSS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR