Anti-HAX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11266-1-AP-150UL
10088-058EA 402 USD
10088-058
Anti-HAX1 Rabbit Polyclonal Antibody
Antibodies
HAX1, also named as HS1BP1, promotes cell survival. It may be involved in internalization of ABC transporters such as ABCB11. HAX1 may inhibit CASP9 and CASP3. It is required to suppress apoptosis in lymphocytes and neurons. This antibody recognize all the isoforms of HAX1.

Western Blot: Human Heart Tissue, 1:200-1:1000; IHC: Human Lymphoma Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: HAX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR