Anti-CD31 Rabbit Polyclonal Antibody

Supplier: Proteintech
11265-1-AP-150UL
10084-326EA 402 USD
10084-326
Anti-CD31 Rabbit Polyclonal Antibody
Antibodies
Platelet endothelial cell adhesion molecule-1 (PECAM-1, CD31) is a member of the immunoglobulin gene superfamily of cell adhesion molecules. It is highly expressed on the surface of the endothelium, making up a large portion of its intracellular junctions. PECAM-1 is also present on the surface of hematopoietic cells and immune cells including platelets, monocytes, neutrophils, natural killer cells, megakaryocytes and some types of T-cell . As well as its role in cell-cell adhesion, PECAM-1 functions as a signaling receptor, and is involved in important physiological events such as nitric oxide production, regulation of T-cell immunity and tolerance, leukocyte transendothelial migration and inflammation and angiogenesis .

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human renal cell carcinoma Tissue, 1:20-1:200; IP:Jurkat Cells, 1:200-1:1000

Type: Primary
Antigen: CD31
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR