Anti-OCT4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11263-1-AP-150UL
10091-540EA 402 USD
10091-540
Anti-OCT4 Rabbit Polyclonal Antibody
Antibodies
Expression of the Oct-4 transcription factor is associated with an undifferentiated phenotype of human and mouse embryonic stem cells and germ cells. Oct-4 (Octamer-4) is a homeodomain transcription factor of the POU family (POU5F1) involved in regulation of pluripotency during normal development and is detectable in embryonic stem (ES) and germ cells. Its expression is associated with an undifferentiated phenotype and tumors. In fact, gene knockdown of Oct4 promotes differentiation, thereby demonstrating a role for this factor in human ES cell self-renewal. As such, it is frequently used as a marker for undifferentiated ES cells or iPS (induced pluripotent stem) cells. Oct4 is one of the four key transcription factors (Oct4, Sox2, c-Myc, and KLF4) used to reprogram mouse and human fibroblasts to a pluripotent state as well as generate patient-specific iPS cells. Affinity purified rabbit anti-Oct4 can be used to demonstrate pluripotency of ES and iPS cells.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: Human embronic stem Cells, 1:20-1:200

Type: Primary
Antigen: OCT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR