Anti-RAD51AP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11255-1-AP-150UL
10093-288EA 402 USD
10093-288
Anti-RAD51AP1 Rabbit Polyclonal Antibody
Antibodies
Homologous recombination (HR) is an important pathway for the elimination of DNA double-strand breaks (DSBs) and other deleterious lesions from chromosomes. The eukaryotic HR reaction is mediated by RAD51 . RAD51AP1 (RAD51 associated protein 1), so named because it can interact with human RAD51, is a vertebrate-specific protein that enhances the ability of RAD51 to mediate the D-loop reaction. RAD51AP1 knockdown by siRNA leads to cellular sensitivity to DNA damaging agents and elevated DNA damage-induced chromatid breaks. Enhancement of the RAD51 recombinase activity by RAD51AP1 is contingent upon physical interaction of the two HR factors .

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: RAD51AP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR