Anti-GLRX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11254-1-AP-150UL
10087-250EA 402 USD
10087-250
Anti-GLRX3 Rabbit Polyclonal Antibody
Antibodies
GLRX3(Glutaredoxin-3) is also named as PICOT, TXNL2. Mammalian glutaredoxin 3 is an essential protein involved in the regulation of signal transduction, for instance during immune cell activation and development of cardiac hypertrophy, presumably in response to redox signals. Glrx3 could take a pivotal role in colon and lung cancer cells during the tumorigenesis, which is expressed in lung and colon cancers consistently and preferentially.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: GLRX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR