Anti-NUDT6 Rabbit Polyclonal Antibody

Supplier: Proteintech
11181-1-AP-150UL
10091-382EA 459.7 USD
10091-382
Anti-NUDT6 Rabbit Polyclonal Antibody
Antibodies
NUDT6, also named as FGF2AS and GFG, belongs to the Nudix hydrolase family. It may contribute to the regulation of cell proliferation. NUDT6 maybe a candidate gene for fat deposition. The 72kd is HomoDimer form of NUDT6.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: NUDT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR