Anti-ABTB1 Rabbit Polyclonal Antibody

11095-1-AP
10082-378EA 356.38 USD
10082-378
Anti-ABTB1 Rabbit Polyclonal Antibody
Proteins with a bric-à-brac, tramtrack, broad-complex/Poxvirus zinc fingers (BTB/POZ) domain are involved in a broad variety of biological processes, including DNA binding, organization of macromolecular structures and regulation of gene transcription . BPOZ contains an N-terminal ankyrin repeat, a bipartite nuclear localization signal and two BTB/POZ domains. It may involve in the development processes .

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: ABTB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR