Anti-PSMB4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11029-1-AP-150UL
10091-880EA 599.76 USD
10091-880
Anti-PSMB4 Rabbit Polyclonal Antibody
Antibodies
PSMB4(Proteasome subunit beta type-4) is also named as PROS26 and belongs to the peptidase T1B family. The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The human protein PSMB4 is known mainly as an interaction partner for several proteins, suggesting an important function of binding not only to the lid, but also to the 20 S core of the proteasome. PSMB4 might be a major site for proteasome regulation, where signals from the outside might be transduced to the protease activities inside. The full length 30 kDa protein has a propeptide of 45 amino acid.

Western Blot: Human Placenta Tissue, 1:200-1:1000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: PSMB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR