Anti-PLOD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11027-1-AP-150UL
10092-484EA 459.7 USD
10092-484
Anti-PLOD3 Rabbit Polyclonal Antibody
Antibodies
PLOD3, also named as LH3, forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links. The major function of PLOD3 in osteoblasts is to glucosylate galactosylhydroxylysine residues in type I collagen.

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IP: HepG2 Cells, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: PLOD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR