Anti-DHX16 Rabbit Polyclonal Antibody

Supplier: Proteintech
11021-1-AP-150UL
10085-832EA 599.76 USD
10085-832
Anti-DHX16 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Lymphoma, 1:10-1:100; IF: MCF-7, 1:20-1:200

Type: Primary
Antigen: DHX16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR