Anti-PPAN Rabbit Polyclonal Antibody

Supplier: Proteintech
11006-1-AP-150UL
10092-620EA 459.7 USD
10092-620
Anti-PPAN Rabbit Polyclonal Antibody
Antibodies
PPAN (or SSF, SSF1 or SSF2) gene is widely expressed, and encodes peter pan homolog protein which may be localized in nucleus. Two isoforms are produced through alternative splicing. So far the characterization of this protein is largely unknown, multiple phosphorylation sites has been revealed based on high-throughput phosphorylation analysis. This antibody is raised against the C-terminal of huaman PPAN and recognize bands around 55 kDa.

Western Blot: Mouse Heart Tissue, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: PPAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR