Anti-HSD17B2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10978-1-AP-150UL
10088-476EA 459.7 USD
10088-476
Anti-HSD17B2 Rabbit Polyclonal Antibody
Antibodies
HSD17B2(Estradiol 17-beta-dehydrogenase 2) is also named as 17-beta-HSD 2,20-alpha-HSD,EDH17B2 and belongs to the short-chain dehydrogenases/reductases (SDR) family.Based on the pivotal role of HSD17B2 gene product in the estrogen metabolism pathway, it may be considered as a putative candidate gene that could possibly explain a fraction of the remaining familial breast cancer risk and it is unlikely to play a role as a high penetrance gene in breast cancer predisposition.

Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: HSD17B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR