Anti-PABPC1,PABP Rabbit Polyclonal Antibody

Supplier: Proteintech
10970-1-AP-150UL
10091-716EA 459.7 USD
10091-716
Anti-PABPC1,PABP Rabbit Polyclonal Antibody
Antibodies
The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation . Polyadenylate-binding protein 1 (PABPC1) is a cytoplasmic-nuclear shuttling protein important for protein translation initiation, and both RNA processing and stability. In the cytoplasm, PABPC1 binds to the 3' poly(A) tail of eukaryotic mRNAs through its RNA-recognition motifs (RRM) and interacts with the N-terminus of eIF4G, part of the eIF4F complex associated with the 5' cap structure .

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human pancreas Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: PABPC1,PABP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR