Anti-FH Rabbit Polyclonal Antibody

Supplier: Proteintech
10966-1-AP-150UL
10082-132EA 459.7 USD
10082-132
Anti-FH Rabbit Polyclonal Antibody
Antibodies
FH(fumarate hydratase) belongs to the class-II fumarase/aspartase family. It is an enzymatic component of the tricarboxylic acid cycle, catalyzing the conversion of fumarate to malate. FH also acts a tumor suppressor functions of FH in sporadic kidney cancer. This protein has 2 isoforms produced by alternative initiation.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: FH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR