Anti-PEX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
10946-1-AP-150UL
10091-460EA 420.09 USD
10091-460
Anti-PEX3 Rabbit Polyclonal Antibody
Antibodies
Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. Peroxin 3, also known as PEX3 (peroxisomal biogenesis factor 3) or TRG18, is a 373 amino acid multi-pass membrane protein that anchored in the peroxisomal membrane and essential for peroxisome biogenesis and integrity. Additionally, Peroxin 3 acts as a docking factor for Peroxin 19 and is required for the import of peroxisomal proteins. Defects in the gene encoding Peroxin 3 are the cause of peroxisome biogenesis disorder complementation group 12 (PBD-CG12) and Zellweger syndrome (ZwS), both of which arise from a failure of peroxisomal protein import.

Type: Primary
Antigen: PEX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR