Anti-NKG2A Rabbit Polyclonal Antibody

Supplier: Proteintech
10935-1-AP-150UL
10091-126EA 459.7 USD
10091-126
Anti-NKG2A Rabbit Polyclonal Antibody
Antibodies
KLRC1, also known as NKG2A (NKG2) and CD159A, is a single-pass type II membrane protein with extracellular C-type lectin domain. It belongs to the natural killer cell lectin-like receptor family, which is a group of transmembrane proteins preferentially expressed in NK cells. KLRC1 can form a heterodimer with CD94. The NKG2A/CD94 complex is an inhibitory receptor that recognizes HLA-E and is expressed by NK cells and a subset of T cells.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: NKG2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR