Anti-MEF2BNB Rabbit Polyclonal Antibody

Supplier: Proteintech
10931-1-AP-150UL
10090-110EA 459.7 USD
10090-110
Anti-MEF2BNB Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:10-1:100

Type: Primary
Antigen: MEF2BNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR