Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSD11B1 Rabbit Polyclonal Antibody

Supplier: Proteintech
HSD11B1(Corticosteroid 11-beta-dehydrogenase isozyme 1) is also named as HSD11, HSD11L,11-DH and 11-beta-HSD1.It is a microsomal enzyme, which acts mainly as a NADP(H)- dependent reductase converting inactive cortisone (E) to active cortisol (F), thus regulating intracellular F access to glucocorticoid receptor. This enzyme is widely expressed not only in liver and adipose tissue, but also in adrenal, ovary and decidua.Western blot analysis in human liver, using a specific11-beta-HSD1 antibody, showed two bands at approximately 34 and 68 kDa confrming the dimeric nature of 11-beta-HSD1 in liver and In omental adipose tissue, an additional band of 50 kDa was strongly expressed.

Western Blot:L02 Cells, 1:200-1:1000

Type: Primary
Antigen: HSD11B1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 10928-1-AP 10088-472 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol HSD11B1
Antigen Name Hydroxysteroid 11-beta Dehydrogenase 1
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms SDR26C1, HSD11B1, 11-beta-hydroxysteroid dehydrogenase 1, MGC13539, hydroxysteroid (11-beta) dehydrogenase 1, HDL, short chain dehydrogenase/reductase family 26C, HSD11, corticosteroid 11-beta-dehydrogenase isozyme 1, 11-DH, member 1, OTTHUMP00000034650, 11-beta-HSD1, HSD11L, HSD11B
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty