Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATP6AP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10926-1-AP
ATP6AP2, also named as ATP6IP2, CAPER, ELDF10, N14F, ATP6M8-9, Renin receptor and prorenin receptor, is believed to potentiate the renin–angiotensin system (RAS), conferring to prorenin, a likely pathological role at tissue level. The PRR has been identified in the microvascular endothelial cells of the retina, in which it seems to be involved in pathological neovascularization processes. The present study demonstrates for the first time that the PRR is expressed in human ATP6AP2 and suggests a molecular mechanism by which hypertension may exacerbate the pathology of dry AMD. ATP6AP2 functions as a renin and prorenin cellular receptor. It may mediate renin-dependent cellular responses by activating ERK1 and ERK2. By increasing the catalytic efficiency of renin in AGT/angiotensinogen conversion to angiotensin I, it may also play a role in the renin-angiotensin system (RAS). Defects in ATP6AP2 are a cause of mental retardation X-linked with epilepsy (MRXE).

Western Blot: Mouse Eye Tissue, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: ATP6AP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 10926-1-AP 10083-358 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ATP6AP2
Antigen Name ATPase H+ transporting accessory protein 2
Reactivity Human, Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen Synonyms MRXSH, PRR, ELDF10, XPDS, HT028, XMRE, MRXE, ATP6M8-9, APT6M8-9, RENR, MSTP009, ATP6IP2, M8-9
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty