Anti-CTCF Rabbit Polyclonal Antibody

Supplier: Proteintech
10915-1-AP-150UL
10085-300EA 459.7 USD
10085-300
Anti-CTCF Rabbit Polyclonal Antibody
Antibodies
Transcriptional insulators are DNA elements that set boundaries on the actions of enhancer and silencer elements and thereby organize the eukaryotic genome into regulatory domains. All vertebrate insulators appear to use the versatile CTCF protein. CTCF uses various combinations of its 11 zinc fingers to recognize a variety of unrelated DNA sequences. Once bound to DNA, CTCF can function as a transcriptional insulator, repressor, or activator, depending on the context of the binding site . In vertebrates, this 11 zinc-finger protein is shown to be crucial in processes of epigenetic imprinting, X chromosome inactivation , and associated with various complex human diseases including cancer and diabetes .

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200; IP: MCF-7 Cells, 1:200-1:1000

Type: Primary
Antigen: CTCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR