Anti-SULT1A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10911-2-AP-150UL
10094-602EA 459.7 USD
10094-602
Anti-SULT1A1 Rabbit Polyclonal Antibody
Antibodies
SULT1A1, also named as ST1A1, P-PST 1, ST1A3, Ts-PST, P-PST 1, STP and STP1, belongs to the sulfotransferase 1 family. It catalyzes the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters. SULT1A1 is also responsible for the sulfation and activation of minoxidil. It mediates the metabolic activation of carcinogenic N-hydroxyarylamines to DNA binding products and could so participate as modulating factor of cancer risk. SULT1A1 is one of two phenol sulfotransferases with thermostable enzyme activity.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200; IP: Mouse Liver Tissue, 1:500-1:5000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: SULT1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR