Anti-EIF3E Rabbit Polyclonal Antibody

Supplier: Proteintech
10899-1-AP-150UL
10082-118EA 459.7 USD
10082-118
Anti-EIF3E Rabbit Polyclonal Antibody
Antibodies
Western Blot:rat lymph Tissue, 1:200-1:2000; IHC: Human pancreas cancer, 1:20-1:200; IF:SH-SY5Y, 1:20-1:200

Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR