Anti-PHF11 Rabbit Polyclonal Antibody

Supplier: Proteintech
10898-1-AP-150UL
10092-184EA 459.7 USD
10092-184
Anti-PHF11 Rabbit Polyclonal Antibody
Antibodies
PHF11 is a transcriptional co-activator of the Th1 effector cytokine genes, interleukin-2 (IL2) and interferon-γ (IFNG), co-operating with nuclear factor kappa B (NF-κB). The reduction in PHF11 expression seen with an AD-associated genotype could contribute to the strong T(H)2 responses that characterize many allergic individuals.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: PHF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR