Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYB5R3 Rabbit Polyclonal Antibody

Supplier: Proteintech
10894-1-AP
CYB5R3, also named as DIA1 and B5R, belongs to the flavoprotein pyridine nucleotide cytochrome reductase family. It plays an essential role in the mitochondrial metabolism by its contribution to cellular redox homeostasis. Overstimulation of CYB5R3 associated with lipid rafts can account for the overshot of plasma membrane-focalized superoxide anion production that triggers the entry of CGN in the irreversible phase of apoptosis.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 10894-1-AP 10085-454 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CYB5R3
Antigen Name Cytochrome b5 reductase 3
Reactivity Human, Rat, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen Synonyms WU:AL591952.1-001, 2500002N19Rik, DIA1, Dia1, WU:Cyb5r3, Dia-1, NADH-cytochrome b5 reductase, C85115, WU:AL591952.1-003, WU:AL591952.1-002, diaphorase 1, Cyb5r3, diaphorase 1 (NADH), 0610016L08Rik
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty