Anti-PLDN Rabbit Polyclonal Antibody

Supplier: Proteintech
10891-2-AP-150UL
10092-444EA 459.7 USD
10092-444
Anti-PLDN Rabbit Polyclonal Antibody
Antibodies
Pallidin, also named as PLDN and PA, is involved in the development of lysosome-related organelles, such as melanosomes and platelet-dense granules. It may play a role in intracellular vesicle trafficking, particularly in the vesicle-docking and fusion process. Pallidin interacts with syntaxin-12 which is a t-SNARE protein that mediates vesicle docking and fusion. Pallidin is a 25 kDa subunit of the octamer BLOC-1.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: PLDN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR