Anti-PTGES2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10881-1-AP-150UL
10093-022EA 459.7 USD
10093-022
Anti-PTGES2 Rabbit Polyclonal Antibody
Antibodies
PTGES2(Prostaglandin E synthase 2) catalyzes the conversion of unstable intermediate of prostaglandin E2 H2 (PGH2) into the more stable prostaglandin E2 (PGE2) form.Its predictedsize is 42 KDa,but it was detected one band of 35KDa in the prostate and two bands of 30KDa,50KDa in liver by western blot. PTGES2 protein is a homotetramer, whereas two PTGES2 molecules each form a strongly interacting dimer.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR