Anti-HSP47 Rabbit Polyclonal Antibody

Supplier: Proteintech
10875-1-AP-150UL
10088-500EA 614.88 USD
10088-500
Anti-HSP47 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HT-1080 Cells, 1:200-1:1000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200; FC: HepG2 Cells, N/A; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: HSP47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR