Anti-ARMET Rabbit Polyclonal Antibody

Supplier: Proteintech
10869-1-AP-150UL
10082-688EA 459.7 USD
10082-688
Anti-ARMET Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ARMET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR