Anti-ELMO2 Rabbit Polyclonal Antibody

10865-1-AP
10086-480EA 349.28 USD
10086-480
Anti-ELMO2 Rabbit Polyclonal Antibody
IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: ELMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR