Anti-LYPLA3 Rabbit Polyclonal Antibody

10863-2-AP
10089-436EA 356.38 USD
10089-436
Anti-LYPLA3 Rabbit Polyclonal Antibody
PLA2G15 is also named as LYPLA3 and belongs to the AB hydrolase superfamily. It has transacylase and calcium-independent phospholipase A2 activity. PLA2G15 catalyzes the formation of 1-O-acyl-N-acetylsphingosine and the concomitant release of a lyso-phospholipid. It can be N-glycosylated.

Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: LYPLA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR