Anti-BLNK Rabbit Polyclonal Antibody

Supplier: Proteintech
10855-1-AP-150UL
10083-628EA 459.7 USD
10083-628
Anti-BLNK Rabbit Polyclonal Antibody
Antibodies
Western Blot: Raji Cells, 1:200-1:1000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: BLNK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR