Anti-SNCA Rabbit Polyclonal Antibody

Supplier: Proteintech
10842-1-AP-150UL
10094-924EA 459.7 USD
10094-924
Anti-SNCA Rabbit Polyclonal Antibody
Antibodies
SNCA, also named as NACP, PARK1 and Alpha-synuclein, belongs to the synuclein family. It may be involved in the regulation of dopamine release and transport. SNCA reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase-3 activation. The deposition of aggregated SNCa is the pathological hallmark of Lewy body disease, which includes Parkinson's disease (PD), diffuse Lewy body disease (DLBD), and Lewy body variant of Alzheimer's disease (LBV). This antibody can recognize all the isoforms of SNCA.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR