Anti-ICAM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10831-1-AP-150UL
10082-152EA 459.7 USD
10082-152
Anti-ICAM1 Rabbit Polyclonal Antibody
Antibodies
ICAM-1 (CD54) is a 90-kDa transmembrane glycoprotein of the immunoglobulin superfamily and is critical for the firm attachment and transmigration of leukocytes out of blood vessels and into tissues. ICAM-1 is expressed by several cell types, typically on endothelial cells and cells of the immune system, and its expression can be up-regulated by various stimuli, including TNF-α, INF-γ, IL-1 and thrombin. It is a ligand for LFA-1 and Mac-1, serves as a receptor for rhinovirus, and is one of several receptors used by Plasmodium falciparum.

Western Blot:L02 Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200; IP: Raji Cells, 1:500-1:5000

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR