Anti-TPT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
10824-1-AP-150UL
10095-906EA 459.7 USD
10095-906
Anti-TPT1 Rabbit Polyclonal Antibody
Antibodies
The translationally controlled tumor protein (TPT1, also known as TCTP) is a highly conserved, abundantly expressed protein associated with cell growth and apoptosis regulation. It is overexpressed in most tumor cells and its down-regulation decreases the viability of those cells. This antibody detected 22-25 kDa TPT1 in lysates. An additional band around 50 kDa can also be observed occasionally, possibly representing the dimer form of TPT1.

Western Blot: Mouse Placenta Tissue, 1:200-1:2000

Type: Primary
Antigen: TPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR