Anti-SESN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10795-1-AP-150UL
10082-224EA 459.7 USD
10082-224
Anti-SESN2 Rabbit Polyclonal Antibody
Antibodies
SESN is a family of highly conserved, stress-inducible genes that can defend the cell against oxidative damage and oncogenic signaling. SESN2 ,which belongs to the SESN family, has been implicated as a tumor suppressor that can inhibit angiogenesis and promote autophagy, underscoring the importance of elucidating the molecular mechanism by which SESN2 regulates pathways of metabolism and survival.. Sesn2 encodes a 480 amino acid protein which can run as a 60 kDa band on SDS-polyacrylamide gel electrophoresis.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: SESN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR