Anti-PRKRA Rabbit Polyclonal Antibody

Supplier: Proteintech
10771-1-AP
10091-730EA 370.14 USD
10091-730
Anti-PRKRA Rabbit Polyclonal Antibody
Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: PRKRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR