Anti-UQCRB Rabbit Polyclonal Antibody

Supplier: Proteintech
10756-1-AP-150UL
10096-582EA 459.7 USD
10096-582
Anti-UQCRB Rabbit Polyclonal Antibody
Antibodies
UQCRB(ubiquinol-cytochrome c reductase binding protein) is also named as UQBP, complex III subunit VII and belongs to the UQCRB/QCR7 family. It is a 13.4 kDa subunit of mitochondrial complex III, plays a crucial role in hypoxia-induced angiogenesis via mitochondrial reactive oxygen species (ROS)-mediated signaling. This protein has 2 isoforms produced by alternative splicing.

Western Blot: Human Heart Tissue, 1:200-1:1000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: UQCRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR