Anti-VCP Rabbit Polyclonal Antibody

Supplier: Proteintech
10736-1-AP-150UL
10096-944EA 459.7 USD
10096-944
Anti-VCP Rabbit Polyclonal Antibody
Antibodies
VCP(Valosin-containing protein) belongs to the AAA ATPase family.VCP was first identified as a result of attempts to clone a putative peptide hormone called valosin. It was found that the cloned cDNA encoded a ubiquitously expressed 90 kDa cytosolic protein, termed VCP, which showed none of the characteristics of a peptide hormone precursor.Defects in VCP are the cause of inclusion body myopathy with early-onset Paget disease and frontotemporal dementia (IBMPFD) and amyotrophic lateral sclerosis type 14 with or without frontotemporal dementia (ALS14).

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: VCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR