Anti-MGP Rabbit Polyclonal Antibody

Supplier: Proteintech
10734-1-AP-150UL
10090-202EA 459.7 USD
10090-202
Anti-MGP Rabbit Polyclonal Antibody
Antibodies
Matrix Gla protein (MGP), a vitamin K-dependent protein, is recognized as a calcification inhibitor in vascular tissue. Defects in MGP are the cause of Keutel syndrome (KS), which is an autosomal recessive disorder characterized by abnormal cartilage calcification, peripheral pulmonary stenosis neural hearing loss and midfacial hypoplasia.

IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: MGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR