Anti-AKR7A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
10730-1-AP-150UL
10082-732EA 459.7 USD
10082-732
Anti-AKR7A2 Rabbit Polyclonal Antibody
Antibodies
AKR7A2(Aflatoxin B1 aldehyde reductase member 2) is also named as AFAR, AFAR1, AKR7 and belongs to the aldo/keto reductase family. AKR7A2 enzyme is catalytically active toward aldehydes arising from lipid peroxidation, suggesting a potential role against the consequences of oxidative stress, and representing an important detoxication route in mammalian cells.

Western Blot: HepG2 Cells, 1:200-1:1000

Type: Primary
Antigen: AKR7A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR